Learn More
Bohlender™ BOLA™ Laboratory Screw Joints EX
Product Code. 10074234
Change view
Click to view available options
Thread Size:
GL 14
GL 18
GL 25
Diameter (Metric) Outer:
4 mm
6 mm
8 mm
10 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size Diameter (Metric) Outer unitSize
10074234 GL 18 6 mm Each
10227644 GL 14 4 mm Each
10402215 GL 18 4 mm Each
10360045 GL 25 8 mm Each
10732037 GL 18 8 mm Each
10594555 GL 18 10 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10074234 Supplier Bohlender™ Supplier No. D84154
Only null left
Add to basket
Add to basket
Laboratory screw joints EX made of conductive PPS-EX with glass fiber and PTFE-EX.
  • Screw cap made of conductive black PPS-EX reinforced with glass fibers
  • Inner parts made of conductive PTFE-EX
Specifications
Material PPS-EX, PTFE-EX
Diameter (English) Outer 0.24 in.
Diameter (Metric) Outer 6 mm
Thread Size GL 18
Color Black
Product Type Screw Joint