missing translation for 'onlineSavingsMsg'
Learn More

[Bis(2-methoxyethyl)amino]sulfur trifluoride, 2.7M (50 wt.%) solution in toluene

Product Code. 10180952 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10180952 25 g Plastic bottle 25g
10531902 5 g Plastic bottle 5g
11955851 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10180952 Supplier Thermo Scientific Acros Supplier No. 368960250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 202289-38-1 | C6H14F3NO2S | 221.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material [Bis(2-methoxyethyl)amino]sulfur trifluoride
CAS 202289-38-1, 108-88-3
CAS Min % 49.0
CAS Max % 51.0
Color Orange-Yellow
Density 1.2000g/mL
Flash Point 4°C
Packaging Plastic bottle
Molecular Formula C6H14F3NO2S
Linear Formula (CH3OCHCH2)2NSF3
Quantity 25 g
Merck Index 15,2899
Specific Gravity 1.2
Synonym bis 2-methoxyethyl aminosulfur trifluoride, deoxo-fluor, 2-methoxy-n-2-methoxyethyl-n-trifluorothio-ethanamine, deoxo-fluor r, bis 2-methoxyethyl aminosulphurtrifluoride, bast, bis 2-methoxyethyl amino sulfur trifluoride, bis 2-methoxyethyl aminosulphur trifluoride
InChI Key APOYTRAZFJURPB-UHFFFAOYSA-N
SMILES COCCN(CCOC)S(F)(F)F
IUPAC Name 2-methoxy-N-(2-methoxyethyl)-N-(trifluoro-$l^{4}-sulfanyl)ethanamine
Molecular Weight (g/mol) 221.24
PubChem CID 2734690
Formula Weight 221.24
Percent Purity ≥95% (NMR, neat substance)
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • ¢Bis(2-methoxyethyl)amino!sulfur trifluoride, 50% solution in toluene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H301-Toxic if swallowed.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P331-Do NOT induce vomiting.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.