missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Falcon™ Biocoat™ 384-Well, Collagen Type I-Treated, Flat-Bottom Microplate

Description
- Source: Rat tail
- TC-treated
- Clear, round well
- Flat bottom
- Non-sterile
- Promotes cell growth, differentiation
- Cell adhesion assays, tissue morphogenesis, cell-matrix interaction, receptor-ligand binding
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Content And Storage | 4°C to 30°C (Stable for 12 months) |
| Array | 16 x 24 |
| Color | White, Clear Bottom |
| Material | Polystyrene |
| No. of Wells | 384 |
| Sterility | Non-sterile |
| Surface Treatment | Collagen Type I |
| Well Shape | Flat Bottom (F-well) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?