missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ BIO-SILICO Silicone Stoppers

Product Code. 10352553
Change view
Click to view available options
Diameter (Metric) Inner:
9-11 mm
13-16 mm
20-23 mm
22-30 mm
30-40 mm
40-50 mm
Unit Size:
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
10352553 9-11 mm Pack of 10
10362553 13-16 mm Pack of 10
10507393 20-23 mm Pack of 10
10776443 22-30 mm Pack of 10
10196060 30-40 mm Pack of 10
10106070 40-50 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10352553 Supplier Hirschmann™ Supplier No. 8905912
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Excellent for the cultivation of aerobic microorganisms. Hirschmann™ SILICOSEN Silicone Stoppers are specially processed culture plugs with coarse bubbles for preparation, filling and sterilization of culture media.

  • Coarse pore structure
  • Good air permeability
  • Chemically and temperature resistant
  • Liquid repellent and reusable

Specifications

Diameter (Metric) Inner 9-11 mm
For Use With (Application) For The Preparation And Sterilization Of Culture Media
Length (Metric) 13 mm
Quantity 10/Pk.
Color Orange
Material Silicone
Product Type Silicone Stopper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.