missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Biebrich Scarlet, dye content approx. 70%

Product Code. 10065680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10065680 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10065680 Supplier Thermo Scientific Chemicals Supplier No. 402221000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4196-99-0 | C22H14N4Na2O7S2 | 556.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 181.0°C to 188.0°C
CAS 4196-99-0
Assay Percent Range 99%
Molecular Formula C22H14N4Na2O7S2
MDL Number MFCD00003891
Synonym Acid Red 66, C.I. 26905, Ponceau BS, Croceine scarlet
Solubility Information Solubility in water: soluble. Other solubilities: soluble in cellosolve, glycol, alcohol
InChI Key YCKMEQRBEVVZQF-VKZXTWAVSA-L
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C(=C1)S([O-])(=O)=O
IUPAC Name disodium 2-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-5-[2-(4-sulfonatophenyl)diazen-1-yl]benzene-1-sulfonate
Molecular Weight (g/mol) 556.47
PubChem CID 16219040
Formula Weight 556.48
Packaging Glass bottle
Quantity 100 g
Chemical Name or Material Biebrich Scarlet, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Biebrich Scarlet
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.