missing translation for 'onlineSavingsMsg'
Learn More

Benzyl mercaptan, 99%

Product Code. 10095980 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10095980 500 g Glass bottle 500g
10239370 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10095980 Supplier Thermo Scientific Acros Supplier No. 421265000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-53-8 | C7H8S | 124.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-53-8
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.20
MDL Number MFCD00004867,MFCD00801588,MFCD01863867
InChI Key UENWRTRMUIOCKN-UHFFFAOYSA-N
Synonym benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan
PubChem CID 7509
IUPAC Name phenylmethanethiol
SMILES SCC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -29°C
Color Colorless to Yellow
Density 1.0580g/mL
Boiling Point 195°C
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2SH
Refractive Index 1.5735 to 1.5775
Quantity 500 g
Beilstein 06, IV, 2632
Merck Index 15, 9475
Specific Gravity 1.058
Solubility Information Solubility in water: insoluble
Formula Weight 124.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Benzyl mercaptan
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.