missing translation for 'onlineSavingsMsg'
Learn More

Benzyl acetate, 99+%

Product Code. 10020550 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10020550 250 mL 250mL
10274700 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10020550 Supplier Thermo Scientific Acros Supplier No. 105822500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 140-11-4 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-11-4
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
InChI Key QUKGYYKBILRGFE-UHFFFAOYSA-N
IUPAC Name benzyl acetate
SMILES CC(=O)OCC1=CC=CC=C1

Specifications

Melting Point -51.0°C
Density 1.0550 g/mL
Boiling Point 214.0°C
Flash Point 102°C
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3CO2CH2C6H5
Refractive Index 1.5010 to 1.5030
Quantity 250 mL
Beilstein 06, 435
Merck Index 15, 1126
Specific Gravity 1.055
Solubility Information Solubility aq. soln.: 1.5 g/L water (20°C). Other solubilities: miscible with alcohol and ether
Formula Weight 150.18
Chemical Name or Material Benzyl acetate
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl acetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.