missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2-Hydroxybenzoate-3,4,5,6-d4, CDN

Product Code. 30322864
Change view
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Product Code. Quantity unitSize
30322864 0.05 g 0.05g
30321908 0.1 g 0.1g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30322864

Brand: CDN CDND67870.05G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl Salicylate-d4
CAS 1219802-40-0
Molecular Formula C14 D4 H8 O3
Quantity 0.05 g
Synonym 6-Hydroxybenzoic-2,3,4,5-d4 acid phenylmethyl ester, 2-Hydroxybenzoic-3,4,5,6-d4 acid benzyl ester, Salicylic-d4 acid benzyl ester, Salicylic Acid Benzyl Ester-d4, Salicylic Acid Benzyl Ester-D4 (salicyl-D4), Benzyl Salicylate-d4, Benzyl 2-Hydroxybenzoate-3,4,5,6-d4
InChI Formula InChI=1 S/C14H12O3/c15-13-9-5-4-8-12(13)14(16)17-10-11-6-2-1-3-7-11/h1-9,15 H,10H2/i4D,5 D,8 D,9 D
SMILES [2 H]c1c([2 H])c([2 H])c(C(=O)OCc2ccccc2)c(O)c1[2 H]
IUPAC Name benzyl 2,3,4,5-tetradeuterio-6-hydroxybenzoate
Molecular Weight (g/mol) 232.2679
Formula Weight 232.1038 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.