missing translation for 'onlineSavingsMsg'
Learn More

Benzophenone imine, 95%, Stab.

Product Code. p-7030532
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15453867 5 g 5g
15463867 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453867 Supplier Thermo Scientific Alfa Aesar Supplier No. H30110.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1013-88-3 | C13H11N | 181.238 g/mol

Useful in the preparation of nitrile ylide dimers.Benzophenone imine as ammonia surrogate. As intermediate material for synthesis of organic chemicals. Catalytic enantioselective alkylation of the benzophenone imine of glycine t-butyl ester using phosphazene (Schwesinger) bases and chiral quaternary ammonium salts yields either enantiomer of amino acid derivatives in excellent yield.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful in the preparation of nitrile ylide dimers.Benzophenone imine as ammonia surrogate. As intermediate material for synthesis of organic chemicals. Catalytic enantioselective alkylation of the benzophenone imine of glycine t-butyl ester using phosphazene (Schwesinger) bases and chiral quaternary ammonium salts yields either enantiomer of amino acid derivatives in excellent yield.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1013-88-3
Molecular Formula C13H11N
Molecular Weight (g/mol) 181.238
MDL Number MFCD00001760
InChI Key SXZIXHOMFPUIRK-UHFFFAOYSA-N
Synonym benzophenone imine, benzophenoneimine, benzhydrylideneamine, 1,1-diphenylmethanimine, unii-ejj21na7vi, benzhydrylimine, benzophenonimine, diphenylmethanimineimine, 1,1-diphenylmethylimine, benzenemethanimine, .alpha.-phenyl
PubChem CID 136809
IUPAC Name diphenylmethanimine
SMILES C1=CC=C(C=C1)C(=N)C2=CC=CC=C2

Specifications

Density 1.08
Boiling Point 151°C to 153°C (10 mmHg)
Refractive Index 1.618
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 181.23
Percent Purity 95%
Chemical Name or Material Benzophenone imine
compliance-icons
Product Identifier
  • Benzophenone imine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Benzophenone imine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.