missing translation for 'onlineSavingsMsg'
Learn More

BD Cell Cocktail

Product Code. 10008763
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10008763 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10008763 Supplier BD Supplier No. 558495

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Three-color reagent cocktail

  • Cocktail component: APC anti-Rat CD3, FITC anti-Rat CD45RA, PE anti-Rat CD161a
  • Components has three different clone: 1F4, OX-33, 10/78 respectively
  • Isotype: mIgM,mIgG1, mIgG1
  • Designed to identify rat T, B, and NK lymphocyte populations by direct immunofluorescent
  • Reactivity: Rat
  • Storage buffer: aqueous buffered solution containing BSA and ≤0.09% sodium azide

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Description APC, FITC, PE
Detectable Analytes Rat T, B and NK lymphocyte populations by direct immunofluorescent
Type Cell Cocktail
For Use With (Application) Flow cytometry
Includes APC Anti-Rat CD3, FITC Anti-Rat CD45RA, PE Anti-Rat CD161a
Quantity 50 Tests
Form Liquid

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.