missing translation for 'onlineSavingsMsg'
Learn More

Batimastat, MedChemExpress

Product Code. 16438437
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. Quantity unitSize
16438437 5 mg 5mg
16435758 10 mg 10mg
2 options
This item is not returnable. View return policy

Product Code. 16438437

Brand: Medchem Express HY13564

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Batimastat is a potent broad spectrum MMP inhibitor with IC50 of 3, 4, 4, 6, and 20 nM for MMP-1, MMP-2, MMP-9, MMP-7 and MMP-3, respectively.

Batimastat is a potent broad spectrum MMP inhibitor with IC50 of 3, 4, 4, 6, and 20 nM for MMP-1, MMP-2, MMP-9, MMP-7 and MMP-3, respectively.

Specifications

Chemical Name or Material Batimastat
Physical Form Solid
CAS 130370-60-4
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C23H31N3O4S2
For Use With (Application) Cancer-Kinase/protease
Synonym BB94
Solubility Information DMSO : 50 mg/mL (104.68 mM; Need ultrasonic)
SMILES O=C(N[C@@H](CC1=CC=CC=C1)C(NC)=O)[C@H](CC(C)C)[C@H](CSC2=CC=CS2)C(NO)=O
Molecular Weight (g/mol) 477.64
Formula Weight 477.64
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.