missing translation for 'onlineSavingsMsg'
Learn More

Barium Nitrate, ACS Reagent, ≥99%, Honeywell Fluka™

Product Code. p-7120215 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15662060 100 g Poly Bottle 100g
15672060 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15662060 Supplier Honeywell-Fluka Supplier No. 217581100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10022-31-8
Molecular Formula BaN2O6
Molecular Weight (g/mol) 261.34
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
Synonym barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2
PubChem CID 24798
IUPAC Name barium(2+);dinitrate
SMILES [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 592°C
Color Colorless
Quantity 100 g
Linear Formula Ba(NO3)2
UN Number UN1446
Formula Weight 261.34g/mol
Percent Purity ≥99%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Crystals
Density 3.24g/cu.cm
Chemical Name or Material Barium Nitrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.