Learn More
Lenz Laborglasinstrumente™ Ball Members, Inch Range
Product Code. 10040201
Change view
Click to view available options
Overall Length (Metric):
120 mm
150 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Overall Length (Metric) unitSize
10040201 150 mm Each
10253951 120 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10040201 Supplier Lenz Laborglasinstrumente™ Supplier No. 01215130
Only null left
Add to basket
Add to basket
Manufactured from DURAN™ tubing. Lenz Laborglasinstrumente™ Ball Members, Inch Range are perfect for mixing solutions in a centrifuge tube.
  • Acc. to DIN 12264 and in addition to standard
  • Made of DURAN™ tubing
Specifications
Certifications/Compliance DIN 12264
Material Borosilicate Glass
Overall Length (Metric) 150 mm
Packaging Quantity 1