missing translation for 'onlineSavingsMsg'
Learn More

AZD3514, MedChemExpress

Product Code. 16466978
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466978 10 mg 10mg
16437056 100 mg 100mg
16433037 50 mg 50mg
16419527 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16466978 Supplier Medchem Express Supplier No. HY16079

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZD3514 is an orally activie and selective androgen receptor (AR) inhibitor. AZD3514 androgen-dependently and -independently inhibits AR signal. AZD351 down-regulates nuclear AR levels in human LNCaP prostate cancer cells in the absence of androgen with an pIC50 value of 5.75. AZD3514 can be used for the research of prostate cancer.

AZD3514 is an orally activie and selective androgen receptor (AR) inhibitor. AZD3514 androgen-dependently and -independently inhibits AR signal. AZD351 down-regulates nuclear AR levels in human LNCaP prostate cancer cells in the absence of androgen with an pIC50 value of 5.75. AZD3514 can be used for the research of prostate cancer.

Specifications

Chemical Name or Material AZD3514
CAS 1240299-33-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H32F3N7O2
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (192.47 mM)
SMILES CC(N1CCN(CCOC2=CC=C(C3CCN(C4=NN5C(CC4)=NN=C5C(F)(F)F)CC3)C=C2)CC1)=O
Molecular Weight (g/mol) 519.56
Formula Weight 519.56
Percent Purity 98.61%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.