missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microspec™ Autoclave Bags
Description
- Clear bags imprinted with blue biohazard labeling designed for safe disposal
- Constructed of thick polypropylene
- Allows for repeated autoclaving
- Strong side seams for securely holding items
- Front write-on panel
Specifications
Specifications
| Content And Storage | Room Temperature |
| Autoclavable | Autoclavable |
| Color | Clear with Blue Print |
| Disposable | Single-use |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Film Description | Polypropylene |
| No. per Case | 200 |
| Outer Packaging | EcoPak |
| Quantity | 1500 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?