Learn More
Eurofins DiscoverX ATR/ATRIP Protein, Active
Description
Serine/threonine-protein kinase ATR activates checkpoint signaling upon genotoxic stresses such as ionizing radiation (IR), ultraviolet light (UV), or DNA replication stalling, thereby acting as a DNA damage sensor. ATR interacts with the ataxia telangiectasia and Rad3 related protein kinase, resulting in its accumulation at intranuclear foci induced by DNA damage.
GenBank NM_001184.3* / GenBank AF451323.1 * Note: the recombinant ATR contains the amino acid substitution M211T with reference toGenBank NM_001184.3. M211T is reported inGenBank XM_001131387.1.
Specifications
Specifications
| Formulation | mg/mL of enzyme in storage buffer. |
| Molecular Weight (g/mol) | 303 kDa (ATR) and 87 kDa (ATRIP) |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | ATR-FLAG-tag / ATRIP-cMyc-tag |
| Expression System | Recombinant enzyme expressed in mammalian cells |
| Protein Form | Full-length |
| Purity or Quality Grade | Combined ≥43% ration 1:1 to 2:1 ATR:ATRIP |
| Protein | ATR/ATRIP, active |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.