missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Aspergillus Monoclonal Antibody (WF-AF-1)
GREENER_CHOICE

Product Code. 10188723
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10188723 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10188723 Supplier Invitrogen™ Supplier No. MA174434

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody specifically recognizes members of the Aspergillus species including A. fumigatus, A. flavus and A. niger, reacting strongly with walls and septae, and to a lesser extent within the cytoplasm of hyphae. This product requires protein digestion pre-treatment of paraffin sections using trypsin or pronase. In Western blot, this antibody detects a band at ~106k Da in Aspergillus fumigatus wall fraction (WF).

Aspergillus species are highly aerobic and are found in almost all oxygen-rich environments, where they commonly grow as molds on the surface of a substrate, as a result of the high oxygen tension. Some Aspergillus species cause serious disease in humans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aspergillus
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone WF-AF-1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Wall fraction of Aspergillus fumigatus.
Purification Method Ammonium Sulfate Precipitation
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Fungi
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.