missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARK5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10150980
Change view
Click to view available options
Quantity:
200 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10150980 200 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10150980 Supplier Invitrogen™ Supplier No. PA515328

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ARK5 Polyclonal Antibody for Western Blot, IHC (P)

NUAK1 (ARK5) is a serine/threonine kinase that plays a role in many cellular functions. The overexpression of this kinase has been associated with tumor invasion and metastasis (colorectal neoplasm). Acts as a regulator of cellular senescence and cellular ploidy by mediating phosphorylation of 'Ser-464' of LATS1, thereby controlling its stability. Controls cell adhesion by regulating activity of the myosin protein phosphatase 1 (PP1) complex. Acts by mediating phosphorylation of PPP1R12A subunit of myosin PP1: phosphorylated PPP1R12A then interacts with 14-3-3, leading to reduced dephosphorylation of myosin MLC2 by myosin PP1. May be involved in DNA damage response: phosphorylates p53/TP53 at 'Ser-15' and 'Ser-392' and is recruited to the CDKN1A/WAF1 promoter to participate in transcription activation by p53/TP53. May also act as a tumor malignancy-associated factor by promoting tumor invasion and metastasis under regulation and phosphorylation by AKT1. Suppresses Fas-induced apoptosis by mediating phosphorylation of CASP6, thereby suppressing the activation of the caspase and the subsequent cleavage of CFLAR. Regulates UV radiation-induced DNA damage response mediated by CDKN1A. In association with STK11, phosphorylates CDKN1A in response to UV radiation and contributes to its degradation which is necessary for optimal DNA repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARK5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene NUAK1
Gene Accession No. O60285, Q641K5
Gene Alias AMP-activated protein kinase family member 5; AMPK-related protein kinase 5; ARK5; AU014801; AW494241; B230104P22Rik; I79_017442; KIAA0537; NUAK family kinase 1; NUAK family SNF1-like kinase 1; NUAK family, SNF1-like kinase, 1; Nuak1; Omphalocele kinase 1; Omphk1; RGD1309956; ZNUAK family, SNF1-like kinase, 1
Gene Symbols NUAK1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 604-634 amino acids from the C-terminal region of human ARK5.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 77976, 9891
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.