missing translation for 'onlineSavingsMsg'
Learn More

Aristolochic acid A, MedChemExpress

Product Code. p-200101136
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
This item is not returnable. View return policy

Product Code. 16433876

Brand: Medchem Express HYN0510

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Aristolochic acid A (Aristolochic acid I; TR 1736) is the main component of plant extract Aristolochic acids, which are found in various herbal plants of genus Aristolochia and Asarum. Aristolochic acid A significantly reduces both activator protein 1 (AP-1) and NF-κB activities. Aristolochic acid A reduces BLCAP gene expression in human cell lines.

Aristolochic acid A (Aristolochic acid I

Specifications

Chemical Name or Material Aristolochic acid A
CAS 313-67-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H11NO7
Quantity 25 mg
Synonym Aristolochic acid I TR 1736
Solubility Information DMSO : 25 mg/mL (73.26 mM; Need ultrasonic)
SMILES OC(C1=CC2=C(OCO2)C3=C4C(C(OC)=CC=C4)=CC([N+]([O-])=O)=C31)=O
Molecular Weight (g/mol) 341.27
Formula Weight 341.27
Percent Purity 98.52%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.