missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Includes
MeltDoctor™ HRM Allele A DNA (20X), 150μL, MeltDoctor™ HRM Allele G DNA (20X), 150μL, MeltDoctor™ HRM Allele A⁄G DNA (20X), 150μL, MeltDoctor™ HRM Primer Mix (20X), 500μL
The kit consists of forward and reverse primers and three DNA templates representing alleles AA, AB, and BB.
Order Info
Shipping Condition: Wet Ice
Recommended Storage
Store at -15° to -25°C.
Specifications
Specifications
| Concentration | 20X |
| Content And Storage | The kit consists of: MeltDoctor™ HRM Allele A DNA (20x), 150 μL, MeltDoctor™ HRM Allele G DNA (20x), 150 μL, MeltDoctor™ HRM Allele A⁄G DNA (20x), 150 μL, MeltDoctor™ HRM Primer Mix (20x), 500 μL Store at -15 °C to -25 °C Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
| Control Template | DNA template representing alleles AA, AB, and BB |
| Format | Tube(s) |
| Gene Symbol | ACTB |
| Kit Type | MeltDoctor HRM Positive Control Kit |
| Label or Dye | High Resolution Melting (HRM) |
| Product Line | MeltDoctor |
| Product Type | HRM Positive Control Kit |
| Quantity | 1 kit |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?