Learn More
Applied Biosystems™ MeltDoctor™ HRM Calibration Plate, 384-well
Shop All Applied Biosystems ProductsDescription
Includes
Contains: 1 plate, 384-well format
Applied Biosystem™ ready-to-use MeltDoctor™ HRM Calibration Plates contain all the components required for pure dye and HRM calibration, reducing the complexity of getting started with HRM experimentation.
- Ready-to-use MeltDoctor HRM Calibration Plates contain all the components required for pure dye and HRM calibration to reduce the complexity of getting started with HRM experimentation
Gene Expression Analysis & Genotyping, Genotyping & Genomic Profiling, High Resolution Melting (HRM) Analysis, Methylation Analysis, Methylation-Sensitive HRM Analysis, PCR & Real-Time PCR, RNAi, Epigenetics & Noncoding RNA Research, Real Time PCR (qPCR)
Order Info
Shipping Condition: Dry Ice
Specifications
Specifications
| Format | 384-well Plate |
| Content And Storage | 1 plate, 384-well format Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
| Calibrated Dye | HRM (High Resolution Melting) Dye |
| For Use With (Equipment) | 7900HT Fast System |
| Product Line | MeltDoctor |
| Product Type | HRM Calibration Plate |
| For Use With (Application) | Real Time PCR (qPCR) |
| Shipping Condition | Dry Ice |
| Quantity | 1 x 384-well plate |
| Plate Compatibility | Calibration Plate |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.