missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rat IgG2a Isotype Control, PE-Texas Red
Description
Rat IgG2a Isotype Control for Flow, Ctrl
Rat IgG2a is an immunoglobulin subclass that is commonly used in research and diagnostic applications. It is produced by the rat immune system and is known for its ability to activate complement and mediate antibody-dependent cell-mediated cytotoxicity (ADCC). Rat IgG2a has a molecular weight of approximately 160 kDa and is composed of two heavy chains and two light chains. It is often used as a control or reference antibody in experiments involving rat models, and is also commonly used in flow cytometry and immunohistochemistry applications.
Specifications
Specifications
| Antigen | Rat IgG2a |
| Applications | Control, Flow Cytometry |
| Classification | Monoclonal |
| Concentration | 100 μg/mL |
| Conjugate | PE-Texas Red |
| Formulation | PBS with 4mg/mL BSA and 0.1% sodium azide |
| Gene Alias | IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 |
| Host Species | Rat |
| Immunogen | Rat IgG2a |
| Purification Method | Purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?