missing translation for 'onlineSavingsMsg'
Learn More

Ras Mouse, Unlabeled, Clone: 18, BD

Product Code. 10074553
Change view
Click to view available options
Quantity:
150 μg
50 μg
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10074553 50 μg Each
15825209 150 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10074553 Supplier BD Biosciences Supplier No. 610001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ras and related proteins of the Ras superfamily play critical roles in the control of normal and neoplastic proliferation. In mammalian cells there are four true Ras proteins (encoded by Ha- ras , N- ras , Ki- ras A, and Ki- ras B ) which, upon mutational activation, can function as independent oncogenes. These proteins relay signals from tyrosine kinases at the plasma membrane which subsequently lead to the nucleus via a network of serine/threonine kinases . The p21ras protein is active in its GTP-bound state. This form is slowly converted to the GDP-bound form by the intrinsic GTPase activity of Ras. This activity is greatly enhanced byGTPase-activating proteins (GAPs) which subsequently lead to removal of the GTP molecule and replacement with GDP. Maintenance of Ras in the GTP form can lead to transformation. One class of Ras mutations, commonly found in human tumors, results in an accumulation of Ras-GTP. The mutant Ras can bind GAP, but GAP bound in this manner seems unable to affect the Ras-GTPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ras
Applications Western Blot
Classification Monoclonal
Clone 18
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Ras (Ha-ras) aa. 1-190
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.