missing translation for 'onlineSavingsMsg'
Learn More

Ly-6G Rat anti-Mouse, Unlabeled, Clone: 1A8, BD

Product Code. 10152954
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10152954 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10152954 Supplier BD Biosciences Supplier No. 551459

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

The 1A8 antibody reacts with Ly-6G, a 21-25kDa GPI-anchored protein. In the bone marrow, Ly-6G is expressed on the majority of the largest cells, which are predominantly granulocytes, and not on lymphoid or erythroid cells. In the periphery, it is expressed on granulocytes. The mAb RB6-8C5 (Cat. No. 557445/553123) recognizes both Ly-6G and Ly-6C and blocks the binding of mAb 1A8 to Ly-6G. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: 1A8
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Ly-6G-transfected EL4J cell line

Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-6G
Applications Flow Cytometry
Classification Monoclonal
Clone 1A8
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Ly-6G-transfected EL4J cell line
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.