missing translation for 'onlineSavingsMsg'
Learn More

I-A(p) Mouse anti-Mouse, FITC, Clone: 7-16.17, BD

Product Code. 10065584
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10065584 0.1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10065584 Supplier BD Biosciences Supplier No. 558921

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The 7-16.17 antibody reacts with the I-A(p) MHC class II alloantigen. It cross-reacts with I-A(b) and with cells from mice of the H-2j, H-2k, H-2q, H-2r, and H-2s haplotypes. Reactivity with other haplotypes (e.g., d, f) has not been observed.

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen I-A(p)
Applications Flow Cytometry
Classification Monoclonal
Clone 7-16.17
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen B10.P mouse spleen, lymph-node, and thymus cells
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.