Learn More
CD62L Mouse anti-Human, FITC, Clone: DREG 56, BD
Description
TheDREG-56 monoclonal antibody specifically binds to CD62L. CD62L is a 76-95 kDa glycoprotein that is also referred to as L-selectin or LECAM-1. CD62L is expressed on neutrophils, monocytes, T- and B-lymphocyte subsets and NK cells. The DREG-56 antibody recognizes the same antigen as LAM-1, and specifically inhibits >90% of binding of human lymphocytes to high endothelial venules (HEV) in frozen sections of peripheral, but not mucosal lymphoid tissue. It thus defines L-selectin as a human lymphocyte homing receptor for peripheral lymph node HEV.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
Specifications
| Antigen | CD62L |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | DREG 56 |
| Conjugate | FITC |
| Description | SELL; L-selectin; LSEL; LAM-1; LECAM-1; LEU8; LNHR; MEL-14; PLNHR; TQ-1 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Supernatant from PMA-activated Human Peripheral Blood Leukocytes |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.