missing translation for 'onlineSavingsMsg'
Learn More

CD24 Rat anti-Mouse, Unlabeled, Clone: J11D, BD

Product Code. 10126514
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10126514 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10126514 Supplier BD Biosciences Supplier No. 553146

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

The J11d antibody reacts with CD24 (Heat Stable Antigen, HSA or HsAg), a variably glycosylated membrane protein found on erythrocytes, granulocytes, monocytes, and lymphocytes. Levels of expression of CD24 vary during differentiation of the T and B cell lineages: Immature B cells in the bone marrow and spleen of adult mice express high levels of CD24, whereas mature peripheral B cells express intermediate levels of CD24. Similarly, the majority of thymocytes express high levels of CD24, while mature thymic and peripheral T cells do not express CD24. Dendritic cells of the thymus, spleen, liver, and epidermal Langerhans cells have also been reported to express CD24. CD24 is not expressed by NK cells. CD24 is a ligand of CD62P (P-selectin). In vitro treatment of splenocytes with J11d antibody plus complement reduces primary IgM responses and proliferative responses to LPS. While the monoclonal antibodies 30-F1 (Cat. no. 558777), M1/69 (Cat. no. 557436), and J11d all react with CD24, they show subtle differences in the level of staining of different cell populations. When possible, investigators should continue to use the same monoclonal antibody as used in previous studies.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD24
Applications Flow Cytometry
Classification Monoclonal
Clone J11D
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Heat Stable Antigen
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen C3H/HeJ mouse thymocytes
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgM κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.