missing translation for 'onlineSavingsMsg'
Learn More

CD144 Rat anti-Mouse, Unlabeled, Clone: 11D4.1, BD

Product Code. 10095434
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10095434 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10095434 Supplier BD Biosciences Supplier No. 555289

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

The 11D4.1 antibody monoclonal antibody specifically binds to mouse CD144, also known as VE-cadherin. CD144 is a type I transmembrane protein and is a member of the cadherin superfamily. VE-cadherin is an endothelial cell-specific, homophilic adhesion molecule. It is concentrated at interendothelial cells contacts and is thought to be involved in the maintenance of cell layer integrity. In vitro and in vivo studies indicate that the 11D.4 mAb interferes with VE-cadherin-mediated intercellular adhesion.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD144
Applications Flow Cytometry
Classification Monoclonal
Clone 11D4.1
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Cdh5; Cadherin-5; CADH5; VE-cadherin; Vascular endothelial cadherin; 7B4
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse VE-Cadherin-Ig Fusion Protein
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.