missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ANKRD20A12P Recombinant Mouse Monoclonal Antibody (K16310_8A11)
GREENER_CHOICE

Product Code. 30827073
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30827073 100 μL
30827381 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827073 Supplier Invitrogen™ Supplier No. MA564442

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: NQVFQQKLLS MKKVQQECEK LEEDKKMLEE EILNLKTHME NSMVELSKLQ EYKSELDERA MQAVEKLEEI HLQEQAQYKK QLEQLNKDII Highest antigen sequence identity to the following orthologs: Mouse - 50%, Pig - 55%, Cynomolgus monkey - 77%.

The ANKRD20A12P gene belongs to the family of ankyrin repeat-containing proteins, which are known for their role in mediating protein-protein interactions through a structural motif called the ankyrin repeat. These motifs are among the most abundant in eukaryotic proteins and play a significant role in various biological processes, including epigenetic and transcriptional regulation. While detailed functional insights specific to ANKRD20A12P are not comprehensively documented, ankyrin repeat proteins generally contribute to cellular architecture, signal transduction, and sometimes even to transcription factor activity modulation. The pseudogene status of ANKRD20A12P indicates that it may not encode a functional protein, but it could still have indirect roles via regulatory mechanisms or as part of genome evolution processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD20A12P
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone K16310_8A11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene ANKRD20A12P
Gene Accession No. Q8NF67
Gene Symbols ANKRD20A12P
Host Species Mouse
Immunogen Recombinant human ANKRD20A12P (aa 171-260).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100874392
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.