missing translation for 'onlineSavingsMsg'
Learn More

Aniline-N,N-d2, CDN

Product Code. 30319224
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
30319224 5 g 5g
30313777 1 g 1g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30319224

Brand: CDN CDND01315G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aniline-N,N-D2
CAS 1122-59-4
Molecular Formula C6 D2 H5 N
Quantity 5 g
Synonym Benzenamine-d2 (9 CI, ACI), Aniline-N,N-d2 (6 CI, 7 CI, 8 CI), N,N-Dideuteroaniline, Aniline-N,N-D2
InChI Formula InChI=1 S/C6H7N/c7-6-4-2-1-3-5-6/h1-5 H,7H2/i/hD2
SMILES [2 H]N([2 H])c1ccccc1
IUPAC Name N,N-dideuterioaniline
Molecular Weight (g/mol) 95.1388
Formula Weight 95.0704 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.