missing translation for 'onlineSavingsMsg'
Learn More

Aniline, ACS, 99+%

Product Code. 11313298
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Unit Size:
1 L
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11313298 1 L 1 L
11323298 100 mL 100mL
11435160 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11313298 Supplier Thermo Scientific Alfa Aesar Supplier No. 036238.K2
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62-53-3 | C6H7N | 93.13 g/mol

It is used to manufacture dyes, photographic chemicals and agricultural chemicals. Its main use is in the manufacture of precursors to polyurethane and other industrial chemicals. Aniline emulsion template is also essential for the synthesis of hollow microspheres of polyaniline. The poly(N-methyl aniline) doped with sulphonic acids plays an important role for humidity sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-53-3
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.13
MDL Number MFCD00007629
InChI Key PAYRUJLWNCNPSJ-UHFFFAOYSA-N
Synonym benzenamine, phenylamine, aminobenzene, aminophen, anilin, kyanol, arylamine, cyanol, benzeneamine, benzidam
PubChem CID 6115
ChEBI CHEBI:17296
IUPAC Name aniline
SMILES NC1=CC=CC=C1

Specifications

Melting Point -6°C
Density 1.021
Boiling Point 183°C to 184°C
Flash Point 70°C (158°F)
Refractive Index 1.586
Quantity 1 L
UN Number UN1547
Beilstein 605631
Merck Index 14,659
Solubility Information Slightly soluble in water.
Formula Weight 93.13
Percent Purity ≥99%
Grade ACS
Physical Form Liquid
Chemical Name or Material Aniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.