missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Androsta-5,16-diene-3beta,17-diol 3-Acetate 17-Trifluoromethanesulfonate, Mikromol™
Description
Androsta-5,16-diene-3beta,17-diol 3-Acetate 17-Trifluoromethanesulfonate, Mikromol
Specifications
Specifications
| Chemical Name or Material | Androsta-5,16-diene-3beta,17-diol 3-Acetate 17-Trifluoromethanesulfonate |
| CAS | 115375-60-5 |
| Analyte or Component Names | Androsta-5,16-diene-3beta,17-diol 3-Acetate 17-Trifluoromethanesulfonate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H29 F3 O5 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | 3β-Acetoxyandrosta-5,16-dien-17-yl trifluoromethanesulfonate, Androsta-5,16-diene-3,17-diol, 3-acetate 17-(trifluoromethanesulfonate), (3β)-, (3β)-Acetoxy-17-(3-trifluoromethylsulfonyloxy)androsta-5,16 diene |
| InChI Formula | InChI=1S/C22H29F3O5S/c1-13(26)29-15-8-10-20(2)14(12-15)4-5-16-17-6-7-19(21(17,3)11-9-18(16)20)30-31(27,28)22(23,24)25/h4,7,15-18H,5-6,8-12H2,1-3H3/t15-,16-,17-,18-,20-,21-/m0/s1 |
| Solution Type | Neat |
| SMILES | CC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CC=C4OS(=O)(=O)C(F)(F)F)[C@@H]3CC=C2C1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?