missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One AMPLIseal™ RT-PCR Adhesive Sealer

Product Code. 10115423
Change view
Click to view available options
Material:
Polypropylene
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
10115423 Polypropylene Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10115423 Supplier Greiner Bio-One Supplier No. 676040
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use in RT-PCR. Greiner Bio-One AMPLIseal™ RT-PCR Adhesive Sealer is a transparent film with minimal autofluorescence.

  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Self-adhesive
  • Acrylate adhesive provides reliable sealing of microplate
  • Minimum evaporation without influencing PCR reactions or fluorescence measurements
TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
Material Polypropylene
Sterility Non-sterile
No. per Case 100
Dimensions (L x W) 141.3 x 79.4 mm
For Use With (Application) RT-PCR
Product Type Microplate Sealer
Adhesive Self
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.