missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfate, 99.5%, for analysis

Product Code. 10184780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Unit Size:
1kg
250g
25kg
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10184780 1 kg Plastic bottle 1kg
10424945 250 g Plastic bottle 250g
10731424 5 kg Plastic bottle 5kg
11904251 25 kg Fibre drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10184780 Supplier Thermo Scientific Acros Supplier No. 205870010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
SMILES N.N.OS(O)(=O)=O

Spécification

Melting Point 280.0°C
Quantity 1 kg
Assay Percent Range 99.5%
Linear Formula (NH4)2SO4
Merck Index 15,552
Solubility Information Solubility in water: 754g/L. Other solubilities: insoluble in alcohol and acetone
IUPAC Name sulfuric acid diamine
Formula Weight 132.14
Percent Purity 99.5%
Grade Analytical
Packaging Plastic bottle
Residue after Ignition 0.01% max. (as SO4)
Density 1.76
Chemical Name or Material Ammonium sulfate
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.