missing translation for 'onlineSavingsMsg'
Learn More

Ammonium dihydrogen phosphate, 99+%, for analysis

Product Code. 10063953 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic Drum
Plastic bottle
Unit Size:
10kg
1kg
2.5kg
250g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10063953 2.5 kg Plastic bottle 2.5kg
10742051 250 g Plastic bottle 250g
10082910 1 kg Plastic bottle 1kg
15199213 10 kg Plastic Drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10063953 Supplier Thermo Scientific Acros Supplier No. undefined
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7722-76-1 | H6NO4P | 115.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
PubChem CID 24402
ChEBI CHEBI:62982
IUPAC Name phosphoric acid amine
SMILES N.OP(O)(O)=O

Specifications

Melting Point 190.0°C
Color White
pH 4 to 4.5 (5% soln. at 20°C)
Physical Form Crystalline Solid
Quantity 2.5 kg
Assay Percent Range 99+%
Linear Formula NH4H2PO4
Merck Index 15,541
Solubility Information Solubility in water: 368g/L (20°C). Other solubilities: slightly soluble in alcohol,practically insoluble in acetone
Formula Weight 115.03
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Density 1.8g/mL
Chemical Name or Material Ammonium dihydrogen phosphate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.