missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Cerium(IV) Nitrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10131800 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10131800 250 g 250g
10650141 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10131800 Supplier Fisher Chemical Supplier No. A/3730/50
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 16774-21-3
Molecular Formula CeH8N8O18
Molecular Weight (g/mol) 548.22
MDL Number 151121
InChI Key WIBGOERAEYJBOT-UHFFFAOYSA-N
IUPAC Name λ⁴-cerium(4+) bis(nitric acid) diamine tetranitrate
SMILES N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 107°C
Color Orange
pH 1
Physical Form Powder/Solid
Quantity 250 g
Formula Weight 548.22g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonium cerium(IV) nitrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising solid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 C
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
  • H272-May intensify fire; oxidiser.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.