missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Ammonia and Ammonium Test Kits

Product Code. 11737315
Change view
Click to view available options
No. of Tests:
90
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Tests unitSize
11737315 90 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11737315 Supplier WTW™ Supplier No. 252072

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Analytes Indophenol blue (Berthelot reaction), NH4-x: Salyticate
For Use With (Application) For waste water, drinking water, sea water
For Use With (Equipment) PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, spectroFlex photometers
Includes Certificates
No. of Tests 90
Sufficient For 25 Tests
Test Method Photometric
Type CombiCheck Ammonium (NH4) cell test kit
compliance-icons
Product Identifier
  • Ammonium Cell Test A6/25 252072_ Method: photometric_0.20 - 8.00 mg/l NH
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : Sodium nitroprusside, troclosene sodium,dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.