missing translation for 'onlineSavingsMsg'
Learn More

Amicarbazone, MedChemExpress

Product Code. 16497346
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497346 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16497346 Supplier Medchem Express Supplier No. HY17513

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Amicarbazone(BAY-MKH3586; BAY314666) is a potent inhibitor of photosynthetic electron transport via binding to the Qb domain of photosystem II (PSII); herbicide with a broad spectrum of weed control.

Amicarbazone(BAY-MKH3586

Specifications

Chemical Name or Material Amicarbazone
Physical Form Solid
CAS 129909-90-6
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C10H19N5O2
Synonym BAY314666 BAY-MKH 3586
Solubility Information H2O : 1 mg/mL (4.14 mM; ultrasonic and heat to 50°C)
SMILES O=C(N1N=C(C(C)C)N(N)C1=O)NC(C)(C)C
Molecular Weight (g/mol) 241.29
Formula Weight 241.29
Percent Purity 99.5%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.