missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AMG 837 hemicalcium salt
GREENER_CHOICE

Product Code. 18769617
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18769617 50 mg 50mg
18718798 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18769617 Supplier Tocris Bioscience™ Supplier No. 6082/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent free fatty acid receptor 1 (FFA1/GPR40) partial agonist

AMG 837 hemicalcium salt is a potent free fatty acid receptor 1 (FFA1/GPR40) partial agonist (EC50 values are 13.5, 22.6 and 31.7 nM at human, mouse and rat receptors, respectively). Interacts allosterically with full FFA1 agonist Docosahexaenoic acid (Cat. No. 3687). Potentiates glucose-dependent stimulation of insulin in vivo. Antidiabetic.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material AMG 837 hemicalcium salt
CAS 1291087-14-3
Synonym (βS)-β-1-Propyn-1-yl-4-[[4'-(trifluoromethyl)[1,1'-biphenyl]-3-yl]methoxy]benzenepropanoic acid hemicalcium salt
Quantity 50 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C26H20F3O3.?Ca
Formula Weight 457.47
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.