missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AMG 837 hemicalcium salt

Description
AMG 837 hemicalcium salt is a potent free fatty acid receptor 1 (FFA1/GPR40) partial agonist (EC50 values are 13.5, 22.6 and 31.7 nM at human, mouse and rat receptors, respectively). Interacts allosterically with full FFA1 agonist Docosahexaenoic acid (Cat. No. 3687). Potentiates glucose-dependent stimulation of insulin in vivo. Antidiabetic.
Specifications
Specifications
| Chemical Name or Material | AMG 837 hemicalcium salt |
| CAS | 1291087-14-3 |
| Synonym | (βS)-β-1-Propyn-1-yl-4-[[4'-(trifluoromethyl)[1,1'-biphenyl]-3-yl]methoxy]benzenepropanoic acid hemicalcium salt |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO |
| Molecular Formula | C26H20F3O3.?Ca |
| Formula Weight | 457.47 |
| Physical Form | Solid |
| Color | White |
| Product Type | Bioactive Small Molecule |
| Show More |
For Research Use Only.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?