missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ TE 70 and TE 70 PWR Semi-Dry Transfer Unit
Shop All Cytiva Products
Click to view available options
Model:
TE 70 ECL
TE 70 PWR
Unit Size:
Each
Description
- Long-lasting, perforated platinum-titanium and stainless-steel electrodes give uniform, contamination-free transfers
- Requires minimal buffer, only enough to saturate blotting papers and membranes
- Uses low current (0.5A) and voltage (30V) for transfers: safer and more economical
- Suitable for gels up to 14 × 16cm and 21 × 26cm
- Tested for use with Amersham™ ECL Western Blotting reagents
- Unique automatic stopping feature; transfer is stopped when buffer gets depleted, preventing overheating
Specifications
Specifications
| Description | ECL Semi-Dry Blotters are designed to deliver outstanding electrophoretic transfer results using low current and voltage with a minimal amount of buffer. Units are available with and without built-in power supplies to give maximum flexibility. |
| Type | Semi-Dry Transfer Unit |
| Gel Dimensions | 14 x 16 cm |
| Includes | 25 sheets of blotting paper, 50 sheets of cellophane, 2 solid masks, masks for 7 x 8 cm and 14 x 16 cm gels. |
| Model | TE 70 PWR |
| Width (Metric) | 38 cm |
| Height (Metric) | 9 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction