missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ambutonium Bromide, Mikromol™
Description
Ambutonium Bromide
Specifications
Specifications
| Chemical Name or Material | Ambutonium Bromide |
| CAS | 115-51-5 |
| Analyte or Component Names | Ambutonium Bromide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H27 N2 O . Br |
| Certifications/Compliance | ISO 17025 |
| Synonym | (3-Carbamoyl-3,3-diphenylpropyl)ethyldimethylammonium bromide (6CI,7CI), Ammonium, (3-carbamoyl-3,3-diphenylpropyl)ethyldimethyl-, bromide (8CI), (3-Carbamyl-3,3-diphenylpropyl)ethyldimethylammonium bromide, 4-Dimethylamino-2,2-diphenylbutyramide ethyl bromide, Aludrox SA, Ambutonium bromide, BL 700, BL 700B, R 100, Benzenepropanaminium, gamma-(aminocarbonyl)-N-ethyl-N,N-dimethyl-gamma-phenyl-, bromide (9CI), Benzenepropanaminium, gamma-(aminocarbonyl)-N-ethyl-N,N-dimethyl-gamma-phenyl-, bromide (1:1) |
| InChI Formula | InChI=1S/C20H26N2O.BrH/c1-4-22(2,3)16-15-20(19(21)23,17-11-7-5-8-12-17)18-13-9-6-10-14-18;/h5-14H,4,15-16H2,1-3H3,(H-,21,23);1H |
| SMILES | [Br-].CC[N+](C)(C)CCC(C(=O)N)(c1ccccc1)c2ccccc2 |
| IUPAC Name | (4-amino-4-oxo-3,3-diphenylbutyl)-ethyl-dimethylazanium;bromide |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?