Learn More
Invitrogen™ UTP Soln. (10 mM)
Description
This Invitrogen ribonucleotide triphosphate (rNTP) solution is a component of the MAXIscript™ Kits (Cat. No. AM1312, AM1316, AM1314, AM1326, AM1324, AM1318, AM1322, AM1320, AM1310, and AM1308) and is supplied in one tube containing 100 μL. Each is subjected to RNase testing as well as a functional assay to ensure high levels of incorporation.
Order Info
Shipping Condition: Dry Ice
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| Concentration | 10 mM |
| For Use With (Application) | In Vitro transcription |
| Label or Dye | Unlabeled |
| Product Line | Ambion |
| Quantity | 100 μL |
| Shipping Condition | Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.