missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hecht Karl™ Assistant™ Amber Glass Bottles
Description
- Amber glass
- Round shoulder
- Ground glass stopper
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Color | Amber |
| Includes | Ground-in Flat Glass Stopper |
| Material | Glass |
| Closure Color | Amber |
| Closure Included | Yes |
| Mouth | Wide |
| Height (Metric) | 128 mm |
| Product Type | Reagent Bottle |
| Closure Material | Glass |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?