Learn More
Bochem™ Aluminum Bosshead
Product Code. 10274171
Change view
Click to view available options
Size (Metric) Grip:
16.5 mm
Unit Size:
Each
Product Code. Size (Metric) Grip unitSize
10274171 16.5 mm Each
1 options
Product Code. 10274171 Supplier Bochem™ Supplier No. 5342
Only null left
Add to basket
Add to basket
Bochem™ aluminum bossheads are used with rods and stand bases to hold and support a range of laboratory apparatus.
  • Aluminum
  • Socket screw M8
  • 90° angle
Specifications
Type Bosshead Clamp
Material Aluminum
Includes M8 Socket Screw
For Use With (Equipment) Rods and Stand Bases
For Use With (Application) Holds and Supports Laboratory Apparatus
Diameter (Metric) 16.5 mm
Size (Metric) Grip 16.5 mm