Learn More
Lenz Laborglasintrumente Alu Rings with Hooks
Product Code. 10109600
Change view
Click to view available options
Size:
NS 19/26 to 24/29
NS 45/40
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
10109600 NS 45/40 Pack of 10
10643311 NS 19/26 to 24/29 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10109600 Supplier Lenz Laborglasintrumente Supplier No. 01102545
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • For cones and sockets to be used with springs
  • For the safe connection of conical ground joints
  • No breakage of glass hooks
Specifications
Quantity 10/Pk.
For Use With (Application) Cones and sockets to be used with springs
Size NS 45/40