missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Reduced contamination risks: AlphaTec™ 37-520 chemical-resistant food handling gloves feature a chlorinated treatment, lessening the risk of product contamination when handling
- Elevated comfort: Thanks to their cotton flocking, these protective gloves are better at absorbing moisture and perspiration, offering enhanced comfort while facilitating donning and doffing
- Enhanced grip: AlphaTec™ 37-520 safety gloves also feature a raised grip surface pattern, improving grip levels, for safer handling
- Specialized protection: In addition, these nitrile gloves guard against potentially hazardous solvents and oil-based chemicals
Specifications
Specifications
| Applications | Industrial |
| Color | Blue |
| Finger Thickness (English) | 13.8 mil |
| Finger Thickness (Metric) | 0.35 mm |
| Latex Free | Yes |
| Material | Nitrile |
| Palm Thickness (English) | 13.8 mil |
| Palm Thickness (Metric) | 0.35 mm |
| Product Type | Chemical-resistant Gloves |
| Size | 9 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?