missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methyl-phenothiazine-10-ethanol, TRC

Product Code. 30476936
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476936 250 mg 250mg
30477831 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30476936 Supplier Toronto Research Chemicals Supplier No. TRCM326370250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha-Methyl-phenothiazine-10-ethanol
CAS 32209-47-5
Molecular Formula C15H15NOS
Quantity 250 mg
Synonym α-Methyl-10H-phenothiazine-10-ethanol, 10-(β-Hydroxypropyl)phenothiazine
InChI Formula InChI=1S/C15H15NOS/c1-11(17)10-16-12-6-2-4-8-14(12)18-15-9-5-3-7-13(15)16/h2-9,11,17H,10H2,1H3
SMILES CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)O
IUPAC Name 1-phenothiazin-10-ylpropan-2-ol
Molecular Weight (g/mol) 257.35
Formula Weight 257.0874
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.