missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ alpha Actinin 4 Polyclonal Antibody

Product Code. 10077332
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10077332 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10077332 Supplier Invitrogen™ Supplier No. 421400

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Rabbit Polyclonal Antibody

alpha Actinin 4 Polyclonal Antibody for Western Blot, IHC, ICC/IF

Alpha actinins belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it is involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments. This gene encodes a nonmuscle, alpha actinin isoform which is concentrated in the cytoplasm, and thought to be involved in metastatic processes. Mutations in this gene have been associated with focal and segmental glomerulosclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha Actinin 4
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Actn4
Gene Accession No. O43707, P57780, Q9QXQ0
Gene Alias actinin alpha 4; ACTININ-4; actinin-Alpha-4; Actn4; alpha actinin 4; alpha-actinin-4; C77391; F-actin cross-linking protein; focal segmental glomerulosclerosis 1; FSGS; FSGS1; non-muscle alpha-actinin 4
Gene Symbols Actn4
Host Species Rabbit
Immunogen Synthetic peptide derived from the N-terminal region of the human, mouse, and orangutan alpha-Actinin-4 protein.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60595, 63836, 81
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.