missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ALDH9A1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10190830
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10190830 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10190830 Supplier Invitrogen™ Supplier No. PA515006

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine and rat based on sequence homology.

ALDH9A1 belongs to the aldehyde dehydrogenase family of proteins. The protein has a high activity for oxidation of gamma-aminobutyraldehyde and other amino aldehydes. The enzyme catalyzes the dehydrogenation of gamma-aminobutyraldehyde to gamma-aminobutyric acid (GABA). This isozyme is a tetramer of identical 54-kD subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH9A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ALDH9A1
Gene Accession No. P49189, Q9JLJ2
Gene Alias 4-trimethylaminobutyraldehyde dehydrogenase; 4-trimethylaminobutyraldehyde dehydrogenase, N-terminally processed; AA139417; Abaldh; aldehyde dehydrogenase (NAD+); aldehyde dehydrogenase 9 family member A1; aldehyde dehydrogenase 9 family, member A1; aldehyde dehydrogenase 9, subfamily A1; aldehyde dehydrogenase 9A1; aldehyde dehydrogenase E3 isozyme; Aldehyde dehydrogenase family 9 member A1; aldehyde dehydrogenase family 9, subfamily A1; ALDH4; ALDH7; ALDH9; Aldh9a1; E3; ESTM40; gamma-aminobutyraldehyde dehydrogenase; gamma-trimethylaminobutyraldehyde dehydrogenase; R-aminobutyraldehyde dehydrogenase; TMABADH; TMABA-DH
Gene Symbols ALDH9A1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 445-477 amino acids from the C-terminal region of human ALDH9A1.
Purification Method Ammonium Sulfate Precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223, 56752
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.